Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020788.1 | NP_001018624.1 | 1 - 2 | 249 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Danio rerio Inventoried Probe spans exons |