Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ela3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024408.1 | NP_001019579.1 | 7 - 8 | 800 | 145 | |
XM_005171867.4 | XP_005171924.1 | 7 - 8 | 868 | 145 | ||
GenBank mRNA | BC095070.1 | - | 7 - 8 | 800 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |