Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025523.1 | NP_001020694.1 | 4 - 5 | 539 | 120 | |
XM_005160740.4 | XP_005160797.1 | 4 - 5 | 1040 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |