Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030178.2 | NP_001025349.2 | 18 - 19 | 5113 | 81 | |
XM_005172621.4 | XP_005172678.1 | 18 - 19 | 5168 | 81 | ||
XM_005172623.4 | XP_005172680.1 | 18 - 19 | 5164 | 81 | ||
XM_021479186.1 | XP_021334861.1 | 18 - 19 | 5076 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |