Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: per1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030183.1 | NP_001025354.1 | 11 - 12 | 1809 | 108 | |
XM_005172625.4 | XP_005172682.1 | 12 - 13 | 2220 | 108 | ||
XM_005172626.4 | XP_005172683.1 | 12 - 13 | 2217 | 108 | ||
XM_005172627.4 | XP_005172684.1 | 12 - 13 | 2220 | 108 | ||
XM_021479187.1 | XP_021334862.1 | 12 - 13 | 2220 | 108 | ||
GenBank mRNA | BC163543.1 | - | 11 - 12 | 1878 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |