Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sat1a.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030199.1 | NP_001025370.1 | 3 - 4 | 363 | 101 | |
GenBank mRNA | BC097197.1 | - | 3 - 4 | 363 | 101 | |
BC171337.1 | - | 3 - 4 | 345 | 101 | ||
BC171340.1 | - | 3 - 4 | 345 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |