Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc24a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030280.1 | NP_001025451.1 | 2 - 3 | 394 | 84 | |
GenBank mRNA | AY538713.1 | - | 2 - 3 | 394 | 84 | |
AY682553.1 | - | 2 - 3 | 394 | 84 | ||
AY682554.1 | - | 2 - 3 | 374 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |