Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itgb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034987.1 | NP_001030159.1 | 2161 | 78 | ||
GenBank mRNA | BC162706.1 | - | 2258 | 78 | ||
DQ149102.1 | - | 2161 | 78 | |||
HQ538840.1 | - | 2431 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio |