Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037436.1 | NP_001032513.1 | 6 - 7 | 1762 | 71 | |
GenBank mRNA | BC109397.1 | - | 6 - 7 | 1762 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |