Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: illr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044742.1 | NP_001038207.1 | 3 - 4 | 289 | 73 | |
GenBank mRNA | AY986757.1 | - | 3 - 4 | 289 | 73 | |
BC163143.1 | - | 3 - 4 | 289 | 73 | ||
BC163144.1 | - | 3 - 4 | 289 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |