Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2b1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044757.1 | NP_001038222.1 | 7 - 8 | 1264 | 65 | |
XM_009300256.3 | XP_009298531.1 | 7 - 8 | 1839 | 65 | ||
XM_021474964.1 | XP_021330639.1 | 7 - 8 | 1345 | 65 | ||
XM_021474965.1 | XP_021330640.1 | 7 - 8 | 1345 | 65 | ||
XM_021474966.1 | XP_021330641.1 | 6 - 7 | 999 | 65 | ||
GenBank mRNA | BC171622.1 | - | 6 - 7 | 1101 | 65 | |
HM449162.1 | - | 6 - 7 | 978 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |