Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfml2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044849.1 | NP_001038314.1 | 2 - 3 | 415 | 83 | |
XM_005160712.4 | XP_005160769.1 | 2 - 3 | 627 | 83 | ||
XM_009294746.3 | XP_009293021.1 | 2 - 3 | 627 | 83 | ||
XM_021468444.1 | XP_021324119.1 | 2 - 3 | 314 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |