Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045037.1 | NP_001038502.1 | 4 - 5 | 422 | 65 | |
XM_005164652.4 | XP_005164709.1 | 4 - 5 | 491 | 65 | ||
XM_005164653.4 | XP_005164710.1 | 4 - 5 | 508 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |