Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045037.1 | NP_001038502.1 | 6 - 7 | 685 | 77 | |
XM_005164652.4 | XP_005164709.1 | 6 - 7 | 754 | 77 | ||
XM_005164653.4 | XP_005164710.1 | 6 - 7 | 771 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |