Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcfc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045064.1 | NP_001038529.1 | 24 - 25 | 4894 | 131 | |
XM_005169304.4 | XP_005169361.1 | 24 - 25 | 4981 | 131 | ||
XM_009306034.3 | XP_009304309.1 | 24 - 25 | 4961 | 131 | ||
XM_009306035.3 | XP_009304310.1 | 24 - 25 | 4954 | 131 | ||
GenBank mRNA | BC139848.1 | - | 24 - 25 | 4899 | 131 | |
BC142544.1 | - | 24 - 25 | 4899 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |