Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076653.1 | NP_001070121.1 | 2 - 3 | 566 | 151 | |
XM_009294243.3 | XP_009292518.1 | 2 - 3 | 666 | 151 | ||
GenBank mRNA | BC124451.1 | - | 2 - 3 | 566 | 151 | |
BC164429.1 | - | 2 - 3 | 508 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |