Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077178.2 | NP_001070646.1 | 12 - 13 | 1644 | 78 | |
XM_005157588.4 | XP_005157645.1 | 12 - 13 | 1445 | 78 | ||
XM_009291665.3 | XP_009289940.1 | 11 - 12 | 1622 | 78 | ||
GenBank mRNA | BC124265.1 | - | 12 - 13 | 1645 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |