Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077178.2 | NP_001070646.1 | 6 - 7 | 999 | 69 | |
XM_005157588.4 | XP_005157645.1 | 6 - 7 | 800 | 69 | ||
XM_009291665.3 | XP_009289940.1 | 5 - 6 | 977 | 69 | ||
XM_009291666.3 | XP_009289941.1 | 6 - 7 | 1006 | 69 | ||
GenBank mRNA | BC124265.1 | - | 6 - 7 | 1000 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |