Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077178.2 | NP_001070646.1 | 9 - 10 | 1370 | 63 | |
XM_005157588.4 | XP_005157645.1 | 9 - 10 | 1171 | 63 | ||
XM_009291665.3 | XP_009289940.1 | 8 - 9 | 1348 | 63 | ||
GenBank mRNA | BC124265.1 | - | 9 - 10 | 1371 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |