Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077250.2 | NP_001070718.2 | 6 - 7 | 994 | 71 | |
GenBank mRNA | BC115335.1 | - | 6 - 7 | 751 | 71 | |
BC124739.1 | - | 6 - 7 | 954 | 71 | ||
EB952759.1 | - | 3 - 4 | 285 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |