Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plod1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077742.2 | NP_001071210.2 | 1 - 2 | 205 | 62 | |
GenBank mRNA | BC125830.1 | - | 1 - 2 | 205 | 62 | |
DQ020093.1 | - | 1 - 2 | 143 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |