Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: creb3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080651.2 | NP_001074120.1 | 6 - 7 | 1191 | 89 | |
XM_021475142.1 | XP_021330817.1 | 5 - 6 | 881 | 89 | ||
GenBank mRNA | BC129318.1 | - | 6 - 7 | 1193 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |