Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081689.1 | NP_001075158.1 | 7 - 8 | 635 | 94 | |
XM_017358460.2 | XP_017213949.1 | 8 - 9 | 759 | 94 | ||
GenBank mRNA | BC072710.1 | - | 7 - 8 | 618 | 94 | |
BC129308.1 | - | 7 - 8 | 635 | 94 | ||
EF591981.1 | - | 6 - 7 | 710 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |