Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082862.1 | NP_001076331.1 | 4 - 5 | 601 | 70 | |
XM_005165050.4 | XP_005165107.1 | 5 - 6 | 881 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Danio rerio Probe spans exons |