Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110450.1 | NP_001103920.1 | 7 - 8 | 974 | 110 | |
GenBank mRNA | BC134941.1 | - | 7 - 8 | 974 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |