Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gli2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130967.1 | NP_571042.1 | 10 - 11 | 1771 | 71 | |
GenBank mRNA | AF085746.2 | - | 10 - 11 | 1771 | 71 | |
AF348156.1 | - | 7 - 8 | 1162 | 71 | ||
BC163542.1 | - | 10 - 11 | 1762 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |