Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131211.1 | NP_571286.1 | 2 - 3 | 924 | 87 | |
XM_005164805.4 | XP_005164862.1 | 3 - 4 | 1025 | 87 | ||
XM_021474926.1 | XP_021330601.1 | 2 - 3 | 1396 | 87 | ||
GenBank mRNA | BC162389.1 | - | 2 - 3 | 809 | 87 | |
BC162401.1 | - | 2 - 3 | 810 | 87 | ||
S80425.1 | - | 2 - 3 | 924 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |