Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271817.1 | NP_001258746.1 | 2 - 3 | 218 | 104 | |
GenBank mRNA | BC059528.1 | - | 2 - 3 | 138 | 104 | |
BC071431.1 | - | 2 - 3 | 170 | 104 | ||
EH478279.1 | - | 2 - 3 | 712 | 104 | ||
EH543119.1 | - | 2 - 3 | 252 | 104 | ||
U66872.1 | - | 1 - 2 | 87 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |