Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131357.1 | NP_571432.1 | 1 - 2 | 219 | 104 | |
GenBank mRNA | AF132084.1 | - | 1 - 2 | 219 | 104 | |
BC162211.1 | - | 1 - 2 | 161 | 104 | ||
BC162544.1 | - | 1 - 2 | 161 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |