Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131357.1 | NP_571432.1 | 3 - 4 | 440 | 84 | |
GenBank mRNA | AF132084.1 | - | 3 - 4 | 440 | 84 | |
BC162211.1 | - | 3 - 4 | 382 | 84 | ||
BC162544.1 | - | 3 - 4 | 382 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |