Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131357.1 | NP_571432.1 | 4 - 5 | 560 | 95 | |
GenBank mRNA | AF132084.1 | - | 4 - 5 | 560 | 95 | |
BC162211.1 | - | 4 - 5 | 502 | 95 | ||
BC162544.1 | - | 4 - 5 | 502 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |