Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131713.2 | NP_571788.2 | 2 - 3 | 407 | 84 | |
GenBank mRNA | BC095063.1 | - | 2 - 3 | 340 | 84 | |
BC164464.1 | - | 1 - 2 | 271 | 84 | ||
CU468784.1 | - | 1 - 2 | 237 | 84 | ||
EE689153.1 | - | 2 - 3 | 464 | 84 | ||
EE697401.1 | - | 1 - 2 | 90 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |