Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131713.2 | NP_571788.2 | 4 - 5 | 742 | 79 | |
GenBank mRNA | BC095063.1 | - | 4 - 5 | 675 | 79 | |
BC164464.1 | - | 3 - 4 | 606 | 79 | ||
CU468784.1 | - | 3 - 4 | 572 | 79 | ||
EE697401.1 | - | 3 - 4 | 425 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |