Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131713.2 | NP_571788.2 | 7 - 8 | 1083 | 65 | |
GenBank mRNA | BC078368.1 | - | 3 - 4 | 375 | 65 | |
BC095063.1 | - | 7 - 8 | 1016 | 65 | ||
BC164464.1 | - | 6 - 7 | 947 | 65 | ||
CU468784.1 | - | 6 - 7 | 913 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |