Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rock2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174863.2 | NP_777288.1 | 20 - 21 | 2783 | 86 | |
GenBank mRNA | AF295804.1 | - | 20 - 21 | 2783 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |