Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183069.1 | NP_898892.1 | 2 - 3 | 258 | 100 | |
GenBank mRNA | BC045296.1 | - | 2 - 3 | 258 | 100 | |
BC165425.1 | - | 2 - 3 | 147 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |