Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194370.1 | NP_919351.1 | 10 - 11 | 1528 | 71 | |
GenBank mRNA | AJ302035.1 | - | 10 - 11 | 1528 | 71 | |
BC081575.1 | - | 10 - 11 | 1589 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |