Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194421.1 | NP_919402.1 | 1 - 2 | 803 | 66 | |
GenBank mRNA | AJ505990.1 | - | 1 - 2 | 874 | 66 | |
AY575769.1 | - | 1 - 2 | 791 | 66 | ||
BC045453.1 | - | 1 - 2 | 709 | 66 | ||
BC065683.1 | - | 1 - 2 | 823 | 66 | ||
DQ812995.1 | - | 1 - 2 | 561 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |