Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rasgef1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199829.1 | NP_956123.1 | 4 - 5 | 606 | 63 | |
XM_005165278.4 | XP_005165335.1 | 5 - 6 | 733 | 63 | ||
XM_005165279.4 | XP_005165336.1 | 4 - 5 | 682 | 63 | ||
XM_005165280.4 | XP_005165337.1 | 3 - 4 | 580 | 63 | ||
GenBank mRNA | BC055678.1 | - | 4 - 5 | 606 | 63 | |
BC075904.1 | - | 3 - 4 | 546 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |