Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199832.2 | NP_956126.1 | 1 - 2 | 155 | 84 | |
GenBank mRNA | AY192987.1 | - | 1 - 2 | 68 | 84 | |
BC044148.1 | - | 1 - 2 | 140 | 84 | ||
BC164324.1 | - | 1 - 2 | 102 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |