Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312897.1 | NP_001299826.1 | 13 - 14 | 2247 | 83 | |
NR_132439.1 | 15 - 16 | 2327 | 83 | |||
GenBank mRNA | BC047798.1 | - | 15 - 16 | 2321 | 83 | |
BC057484.1 | - | 12 - 13 | 1585 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |