Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312897.1 | NP_001299826.1 | 1 - 2 | 361 | 94 | |
NR_132439.1 | 2 - 3 | 231 | 94 | |||
GenBank mRNA | AW153313.1 | - | 1 - 2 | 343 | 94 | |
BC047798.1 | - | 2 - 3 | 231 | 94 | ||
BC057484.1 | - | 1 - 2 | 312 | 94 | ||
BI876598.1 | - | 2 - 3 | 221 | 94 | ||
BM104529.1 | - | 1 - 2 | 299 | 94 | ||
BM776396.1 | - | 1 - 2 | 353 | 94 | ||
BQ616560.1 | - | 1 - 2 | 310 | 94 | ||
CK689483.1 | - | 1 - 2 | 265 | 94 | ||
CO957401.1 | - | 2 - 3 | 264 | 94 | ||
DT878768.1 | - | 1 - 2 | 353 | 94 | ||
HM151010.1 | - | 1 - 2 | 170 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |