Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312897.1 | NP_001299826.1 | 2 - 3 | 409 | 88 | |
NR_132439.1 | 3 - 4 | 279 | 88 | |||
GenBank mRNA | AW153313.1 | - | 2 - 3 | 391 | 88 | |
BC047798.1 | - | 3 - 4 | 279 | 88 | ||
BC057484.1 | - | 2 - 3 | 360 | 88 | ||
BI876598.1 | - | 3 - 4 | 269 | 88 | ||
BM104529.1 | - | 2 - 3 | 347 | 88 | ||
BM776396.1 | - | 2 - 3 | 401 | 88 | ||
BQ616560.1 | - | 2 - 3 | 358 | 88 | ||
CK689483.1 | - | 2 - 3 | 313 | 88 | ||
CO957401.1 | - | 3 - 4 | 312 | 88 | ||
DT878768.1 | - | 2 - 3 | 401 | 88 | ||
HM151010.1 | - | 2 - 3 | 218 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |