Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199836.2 | NP_956130.1 | 3 - 4 | 417 | 109 | |
GenBank mRNA | BC044149.1 | - | 3 - 4 | 418 | 109 | |
BC163944.1 | - | 3 - 4 | 261 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |