Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328344.1 | NP_001315273.1 | 3 - 4 | 348 | 87 | |
GenBank mRNA | BC046008.1 | - | 3 - 4 | 281 | 87 | |
EH488960.1 | - | 3 - 4 | 346 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |