Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200247.1 | NP_956541.1 | 2 - 3 | 416 | 117 | |
GenBank mRNA | BC047831.1 | - | 2 - 3 | 416 | 117 | |
BC164589.1 | - | 1 - 2 | 307 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |