Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: maco1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200463.2 | NP_956757.2 | 6 - 7 | 1380 | 69 | |
XM_005157048.4 | XP_005157105.1 | 6 - 7 | 1356 | 69 | ||
XM_021480543.1 | XP_021336218.1 | 6 - 7 | 1461 | 69 | ||
GenBank mRNA | AY845030.1 | - | 6 - 7 | 1380 | 69 | |
BC171392.1 | - | 6 - 7 | 1350 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |