Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200660.1 | NP_956954.1 | 6 - 7 | 746 | 63 | |
GenBank mRNA | BC057508.1 | - | 6 - 7 | 746 | 63 | |
BC164095.1 | - | 6 - 7 | 759 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |