Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200786.1 | NP_957080.1 | 7 - 8 | 1036 | 75 | |
XM_009305682.2 | XP_009303957.1 | 7 - 8 | 1061 | 75 | ||
GenBank mRNA | BC059635.1 | - | 7 - 8 | 1036 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |