Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200865.2 | NP_957159.1 | 8 - 9 | 749 | 141 | |
GenBank mRNA | BC063230.1 | - | 8 - 9 | 683 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |